Please login to your online account to display your discounted pricing

Molecular Sieve, Activated, Type 5A, 8 to 12 Mesh, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 250g
Packaging HDPE Pressure Bottom Poly Bottle

Manufacturer: avantor performance  270903

 View more versions of this product

Catalog No. 02003413

  • / Case of 12 Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Molecular Sieve
CAS 1344-01-0
Name Note Activated, Type 5A
Purity Grade BAKER ANALYZED™ Reagent
Mesh Size 8 to 12 Mesh
Packaging HDPE Pressure Bottom Poly Bottle
Specific Gravity 1L = 2.10kg
Quantity 250g

Specifications

Chemical Name or Material Molecular Sieve
CAS 1344-01-0
Name Note Activated, Type 5A
Purity Grade BAKER ANALYZED™ Reagent