Please login to your online account to display your discounted pricing

Molecular Sieve, Activated, Type 4A, 8 to 12 Mesh, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Wide Mouth Amber Glass Bottle

Manufacturer: avantor performance  270801

 View more versions of this product

Catalog No. 02003414

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Molecular Sieve
CAS 1344-01-0
Name Note Activated, Type 4A
Purity Grade BAKER ANALYZED™ Reagent
Mesh Size 8 to 12 Mesh
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.10kg
Quantity 500g

Specifications

Chemical Name or Material Molecular Sieve
CAS 1344-01-0
Name Note Activated, Type 4A
Purity Grade BAKER ANALYZED™ Reagent