Please login to your online account to display your discounted pricing

Molecular Sieve, Activated, Type 4A, 8 to 12 Mesh, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$167.33 - $711.20

Chemical Identifiers

Chemical Name or Material Molecular Sieve
CAS 1344-01-0
Name Note Activated, Type 4A
Purity Grade BAKER ANALYZED™ Reagent
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003414

 
avantor performance
270801
Wide Mouth Amber Glass Bottle 500g Each for $167.33

02003417

 
avantor performance
270805
Wide Mouth PVC Coated Glass Bottle 2.5kg Each for $711.20
Description & Specifications

Specifications

Chemical Name or Material Molecular Sieve
CAS 1344-01-0
Purity Grade BAKER ANALYZED™ Reagent
Quantity 500g
Mesh Size 8 to 12 Mesh
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.10kg
Name Note Activated, Type 4A