missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Molecular Sieve, Activated, Type 3A, 8 to 12 Mesh, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Supplier: Avantor 271001
Specifications
8 to 12 Mesh | |
1L = 2.10kg | |
Molecular Sieve | |
1344-01-0 | |
IDZYHGDLWGVHQM-UHFFFAOYSA-N | |
aluminum;calcium;sodium;silicate | |
44144394 |
500 g | |
Wide Mouth Amber Glass Bottle | |
Activated, Type 3A | |
AlCaNaO4Si+2 | |
[O-][Si]([O-])([O-])[O-].[Na+].[Al+3].[Ca+2] | |
182.13 | |
BAKER ANALYZED™ Reagent |
Chemical Identifiers
1344-01-0 | |
182.13 | |
44144394 | |
[O-][Si]([O-])([O-])[O-].[Na+].[Al+3].[Ca+2] |
AlCaNaO4Si+2 | |
IDZYHGDLWGVHQM-UHFFFAOYSA-N | |
aluminum;calcium;sodium;silicate |