missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Molecular Probes™ TSA™ Kit 3, with HRP-Goat Anti-Mouse IgG and Alexa Fluor™ 546 Tyramide DFS Item

Tyramide signal amplification is enzyme-mediated detection method that uses catalytic activity of horseradish peroxidase (HRP) to generate high-density labeling of target protein or nucleic acid sequence in situ

Manufacturer:  Invitrogen™ LST20913

Catalog No. T20913



Includes: HRP-goat anti-mouse IgG and orange-fluorescent Alexa Fluor 546 tyramide

Description

Description

  • Alexa Fluor 546 tyramide is spectrally similar to Cy3 dye

Cell Analysis, Cellular Imaging, Fluorescence In Situ Hybridization, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Staining and Detection, Immunohistochemistry (IHC), In Situ Hybridization (ISH)

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Store in freezer (-5° to -30°C) and protect from light.
Documents

For Research Use Only. Not for use in diagnostic procedures.