Please login to your online account to display your discounted pricing

Molecular Probes™ TetraSpeck™ Fluorescent Microspheres Size Kit (mounted on slide) DFS Item

For calibration of wide field, TIRF and confocal fluorescence microscopes, especially for multicolor applications like colocalization analysis and derivation of point spread functions critical for deconvolution

$372.00 - $372.00

Products
Catalog Number Mfr. No. Price Quantity    

T14792

DFS Item
 
life technologies
LST14792
Each for $372.00
Description & Specifications

  • Contains one microscope slide, with six viewing regions
  • Each of five viewing regions contains microspheres of one size: 0.1, 0.2, 0.5, 1.0 or 4.0μm
  • Remaining region contains mixture of all five bead sizes
  • Every microsphere is stained with four different fluorescent dyes [365/430nm (blue), 505/515nm (green), 560/580nm (orange), and 66/680nm (dark red)] that have well-separated excitation and emission peaks

Cell Analysis, Cell Cycle, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF)

Shipping Condition: Room temperature