Please login to your online account to display your discounted pricing

Molecular Probes™ SYPRO™ Red Protein Gel Stain (5,000X Concentrate in DMSO) DFS Item

Fast, one-step staining protocol requiring no destaining

$300.00 - $339.00

Specifications

For Use With (Application) Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Storage Requirements Store at room temperature and protect from light.
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

S6653

DFS Item
 
life technologies
LSS6653
500μL Each for $300.00

S6654

DFS Item
 
life technologies
LSS6654
10 × 50μL Each for $339.00
Description & Specifications

Specifications

For Use With (Application) Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Storage Requirements Store at room temperature and protect from light.

SYPRO Red protein gel stain is a sensitive, ready-to-use fluorescent stain for proteins in 1-D gels. It offers many advantages over Coomassie Blue staining, including a linear dynamic range over three orders of magnitude and very little protein-to-protein variation in staining. Stained proteins can be viewed with a standard UV or blue-light transilluminator or with a laser scanner.

Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Shipping Condition: Room Temperature