missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ SYPRO™ Orange Protein Gel Stain (5,000X Concentrate in DMSO)
A sensitive, ready-to-use fluorescent stain for total protein detection in 1D gels with a one-step protocol that does not require destaining.
$382.00 - $417.00
Specifications
Concentration | 5000X |
---|---|
Detection Location | In-Gel Detection |
Detection Method | Fluorescent |
Excitation/Emission | 300, 470/570 nm |
Product Line | SYPRO™ |
Includes: Supplied as a 5000X concentrate in DMSO.
Description
- A sensitive, ready-to-use fluorescent stain for total protein detection in 1D gels
- The sensitivity of SYPRO Orange Protein Stain is as good as, or better than traditional silver staining techniques
- Stained proteins can be viewed with a standard UV, blue-light transilluminator, or imaging equipment containing the appropriate filters/lasers
- Fast, one-step staining protocol requiring no destaining
- Linear quantitation range over three orders of magnitude
- Very little protein-to-protein variation in staining
- Store at room temperature and protect from light
Order Info
Shipping Condition: Room Temperature
Specifications
5000X | |
Fluorescent | |
SYPRO™ |
In-Gel Detection | |
300, 470/570 nm | |
Protein |
For Research Use Only. Not for use in diagnostic procedures.