Please login to your online account to display your discounted pricing

Molecular Probes™ SNARF™ 5-(and-6)-Carboxy SNARF -1, Acetoxymethyl Ester, Acetate DFS Item


Cell-permeant pH indicator has pKa of approximately 7.5 after de-esterification, thus is useful for measuring pH changes between pH 7 and pH 8

Manufacturer: life technologies  LSC1271

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. C1271

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) Flow Cytometer, Fluorescence Microscope, Microplate Reader
Storage Requirements Store in freezer (-5 to -30°C).
Quantity 1mg