Please login to your online account to display your discounted pricing

Molecular Probes™ SNAP (S-Nitroso-N-Acetylpenicillamine) DFS Item


Useful for cell viability, proliferation and function

Manufacturer: life technologies  LSN7892

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. N7892

  • / Each
 

Description & Specifications

Specifications

Sensor Types Free Radical Generator
Detection Method Fluorescent Dye-Based
Storage Conditions Store in freezer (-5° to -30°C) and protect from light.
Quantity 25mg