missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molecular Probes™ SNAP (S-Nitroso-N-Acetylpenicillamine) DFS Item

Useful for cell viability, proliferation and function

$128.10 - $128.10

Specifications

Storage Conditions Store in freezer (-5° to -30°C) and protect from light.
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N7892 Life Technologies
LSN7892
25mg Each for $128.10
Specifications

Description

  • SNAP spontaneously releases nitric oxide (NO) under physiological conditions

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Nitric Oxide Research, Nitro-Oxidative Stress

Order Info

Shipping Condition: Room Temperature

Description & Specifications

Specifications

Store in freezer (-5° to -30°C) and protect from light.