Please login to your online account to display your discounted pricing

Molecular Probes™ SIN-1 (3-Morpholinosydnonimine, Hydrochloride) DFS Item

Useful for cell viability, proliferation and function

$164.85 - $164.85

Specifications

Storage Conditions Store in freezer (-5° to -30°C) and protect from light.
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

M7926

DFS Item
 
life technologies
M7926
20 x 1mg Each for $164.85
Description & Specifications

Specifications

Storage Conditions Store in freezer (-5° to -30°C) and protect from light.

  • SIN-1 spontaneously releases nitric oxide (NO) and superoxide under physiological conditions

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Nitric Oxide Research, Nitro-Oxidative Stress

Shipping Condition: Room Temperature