Please login to your online account to display your discounted pricing
Molecular Probes™ SIN-1 (3-Morpholinosydnonimine, Hydrochloride) DFS Item
Useful for cell viability, proliferation and function
Manufacturer: life technologies  M7926
 View more versions of this product
Catalog No. M7926
  • / Each
Description & Specifications
Specifications
Quantity 20 x 1mg
Storage Conditions Store in freezer (-5° to -30°C) and protect from light.
Sensor Types Free Radical Generator
  • SIN-1 spontaneously releases nitric oxide (NO) and superoxide under physiological conditions
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Nitric Oxide Research, Nitro-Oxidative Stress