Please login to your online account to display your discounted pricing

Molecular Probes™ PRO-Q™ Multiplexed Proteomics Phosphoprotein Gel Staining Kit DFS Item


For distinguishing a low amount of a highly phosphorlyated protein from a higher amount of a less phosphorylated protein

Manufacturer: life technologies  LSMPM33305

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. MPM33305

  • / Each
 

Description & Specifications

Specifications

Kit Contents MPP33300, 40uL Standard and S12000
For Use With (Application) Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Color Ruby