Molecular Probes™ PRO-Q™ Multiplexed Proteomics Phosphoprotein Gel Staining Kit DFS Item
For distinguishing a low amount of a highly phosphorlyated protein from a higher amount of a less phosphorylated protein
$329.00 - $1,136.00
Specifications
For Use With (Application) Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
View More Specs
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Products ${productFamilyLength}
Catalog Number Mfr. No. Kit Contents Price Quantity  
Catalog Number Mfr. No. Kit Contents Price Quantity  
MPM33305 Life Technologies
LSMPM33305
MPP33300, 40uL Standard and S12000 Each for $1,136.00
MPM33306 Life Technologies
LSMPM33306
MPP33301, 40uL Standard and S12001 Each for $329.00
Specifications
Description
The Multiplexed Proteomics™ Phosphoprotein Gel Stain Kit contains the Pro-Q™ Diamond phosphoprotein gel stain and the SYPRO™ Ruby protein gel stain. Determining the ratio of the Pro-Q Diamond dye to the SYPRO Ruby dye signal intensities for each band or spot provides a measure of the phosphorylation level normalized to the total amount of protein. Using both stains in combination makes it possible to distinguish a low amount of a highly phosphorlyated protein from a higher amount of a less phosphorylated protein.
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Description & Specifications
Specifications
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis