Molecular Probes™ PRO-Q™ Diamond Phosphoprotein Gel Staining Kit DFS Item
Can detect as little as 1-16ng of phosphoprotein per band with a linear signal over three orders of magnitude
$217.00 - $2,510.00
Specifications
For Use With (Application) Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
View More Specs
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Products ${productFamilyLength}
Catalog Number Mfr. No. Kit Contents Price Quantity  
Catalog Number Mfr. No. Kit Contents Price Quantity  
MPP33300 Life Technologies
LSMPP33300
1L Stain, 40 μL Standard Each for $638.00
MPP33301 Life Technologies
LSMPP33301
200mL of the Pro-Q™ Diamond Phosphoprotein Gel Stain and 40μL of the PeppermintStick™ phosphoprotein molecular weight standard Each for $217.00
MPP33302 Life Technologies
LSMPP33302
5L of the Pro-Q™ Diamond Phosphoprotein Gel Stain and 40μL of the PeppermintStick™ phosphoprotein molecular weight standards Each for $2,510.00
Specifications
Description
The Pro-Q Diamond Phosphoprotein Gel Staining Kit, includes 1L of the Pro-Q Diamond Phosphoprotein Gel Stain and 40μL of the PeppermintStick™ phosphoprotein molecular weight standards. The PeppermintSTick™ phosphoprotein molecular weight standards are a mixture of phosphorylated and nonphosphorylated proteins with molecular weights from 14,000 to 116,250 daltons that serve both as molecular weight markers and as positive and negative controls.
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Description & Specifications
Specifications
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis