Please login to your online account to display your discounted pricing
Molecular Probes™ PRO-Q™ Diamond Phosphoprotein Gel Staining Kit DFS Item
Can detect as little as 1-16ng of phosphoprotein per band with a linear signal over three orders of magnitude
Manufacturer: life technologies  LSMPP33302
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. MPP33302
  • / Each
Description & Specifications
Specifications
Kit Contents 5L of the Pro-Q™ Diamond Phosphoprotein Gel Stain and 40μL of the PeppermintStick™ phosphoprotein molecular weight standards
For Use With (Application) Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis