Molecular Probes™ Press-to-Seal™ Silicone Sheet DFS Item
For versatility and flexibility, isolators can be easily trimmed to custom fit enclosures
$226.00 - $298.00
Specifications
Dimensions (L x W) 13 x 18cm
View More Specs
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
P18178 Life Technologies
LSP18178
0.5mm thick silicon sheet Each for $226.00
P18179 Life Technologies
LSP18179
1.0mm thick silicon sheet Each for $298.00
Specifications
Description
  • Available either precut to specific interior dimensions or as sheet material that can be trimmed
  • Isolators are autoclavable and non-cytotoxic
  • Can be used to isolate cells in culture dishes or to separate specimens on microscope slides
Cell Analysis, Cellular Imaging, IHC Sample Preparation, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunohistochemistry (IHC)
Order Info
Shipping Condition: Room temperature
Description & Specifications
Specifications
13 x 18cm