Molecular Probes™ Press-to-Seal™ Silicone Isolator DFS Item
For versatility and flexibility, isolators can be easily trimmed to custom fit enclosures
$120.75 - $238.00
Specifications
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Wells Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Wells Price Quantity  
P18174 Life Technologies
LSP18174
20mm 1 well, 20mm dia., 0.5mm deep Each for $181.00
P18175 Life Technologies
LSP18175
20mm 1 well, 20mm dia., 1.0mm deep Each for $181.00
P24741 Life Technologies
LSP24741
20mm 1 well, 20mm dia., 1.0mm deep Each for $120.75
P24743 Life Technologies
LSP24743
9mm 8 wells, 9mm dia., 0.5mm deep Each for $172.00
P24742 Life Technologies
LSP24742
2.5mm 24 wells, 2.0mm dia., 2.0mm deep Each for $238.00
P24744 Life Technologies
LSP24744
9mm 8 wells, 9mm dia., 1.0mm deep Each for $170.00
Specifications
Description
  • Available either precut to specific interior dimensions
  • Isolators are autoclavable and noncytotoxic
  • Can be used to isolate cells in culture dishes or to separate specimens on microscope slides
Cell Analysis, Cellular Imaging, IHC Sample Preparation, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunohistochemistry (IHC)
Order Info
Shipping Condition: Room temperature
Description & Specifications
Specifications
0.0