Invitrogen™ Molecular Probes™ Press-to-Seal™ Silicone Isolator DFS Item
For versatility and flexibility, isolators can be easily trimmed to custom fit enclosures
Manufacturer:  Invitrogen™ LSP24741
 View more versions of this product
Catalog No. P24741
Specifications
Specifications
Press-to-Seal™ Silicone Sheet with Adhesive
1 well, 20mm dia., 1.0mm deep
20mm
Documents
For Research Use Only. Not for use in diagnostic procedures.