Molecular Probes™ Premo™ FUCCI Cell Cycle Sensor (BacMam 2.0) DFS Item
Ready-to-use format
Manufacturer:  Life Technologies LSP36237
 View more versions of this product
Catalog No. P36237
Disclaimers:
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
Specifications Description & Specifications
Specifications
2 x 1000μL
Fluorescent
Cell Cycle Sensor
GFP (EmGFP), RFP (TagRFP)
Store at 2° to 6°C, protect from light. Do not freeze.
Flow Cytometer, Fluorescence Microscope, Fluorescent Imager, High Content Instrument, Tali™ Image-Based Cytometer
488/510 (Green), 555/584 (Red)
Green, Red-Orange