Invitrogen™ Molecular Probes™ NBD C12-HPC (2-(12-(7-Nitrobenz-2-Oxa-1,3-Diazol-4-yl)amino)Dodecanoyl-1-Hexadecanoyl-sn-Glycero-3-Phosphocholine) DFS Item
Fluorescent-labeled phospholipid
Manufacturer:  Invitrogen™ LSN3787
Catalog No. N3787
Description
Description
The phospholipid, NBD C12-HPC is labeled on the acyl chain with the environment sensitive fluorophore, NBD (excitation/emission maxima approx. 463/536nm).
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining and Detection
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
5mg
Store in freezer (-5° to -30°C) and protect from light.
Fluorescent Labeled Phospholipid
SDS
Documents
For Research Use Only. Not for use in diagnostic procedures.