Please login to your online account to display your discounted pricing

Molecular Probes™ Mag-Fura-2, AM, Cell Permeant DFS Item


Intracellular magnesium indicator that is ratiometric and UV light-excitable

Manufacturer: life technologies  LSM1291

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. M1291

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) Flow Cytometer, Fluorescence Microscope, Microphotometer
Storage Requirements Store in freezer (-5° to -30°C) and protect from light.
Quantity 1mg