Learn More
Invitrogen™ Image-IT™ LIVE Intracellular Membrane and Nuclear Labeling Kit (Counterstains For GFP-Expressing Cells)
Counterstains for GFP-expressing cells
Supplier: Invitrogen™ I34407
Description
The Image-iT LIVE Intracellular Membrane and Nuclear Labeling Kit provides two stains - red-fluorescent CellTrace™ BODIPY™ TR methyl ester and blue-fluorescent Hoechst 33342 dye - for highly selective staining of the intracellular membranes and nuclei of live green-fluorescent protein (GFP)-transfected cells and tissues.
Cell Analysis, Cell Structure, Cell-Based Reporter Assays, Cellular Imaging, Enzyme & Protein Activity Assays, Fluorescent Protein (e.g. GFP) Assays, Fluorescent Protein Assays (GFP), Immunofluorescence (IF), Immunofluorescence Staining & Detection, Nucleus, Nucleoli & Nuclear Envelope, Plasma Membrane, Proteins, Expression, Isolation & Analysis, Reporter Assays
Order Info
Shipping Condition: Room Temperature
Specifications
Red, Blue | |
Fluorescence Microscope | |
Image-iT™ | |
Labeling Kit |
Store in freezer -5°C to -30°C and protect from light. | |
Fluorescent | |
Room Temperature | |
Nucleus, Cell Membranes & Lipids |
For Research Use Only. Not for use in diagnostic procedures.