missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Hoechst 33342, Trihydrochloride, Trihydrate, 100 mg DFS Item

Nucleic acid stain

$100.00 - $133.00

Specifications

Detection Method Fluorescent
For Use With (Equipment) Fluorescence Microscope
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H3570 View Documents Invitrogen™
LSH3570
10mL Each for $133.00
 
H1399 View Documents Invitrogen™
LSH1399
100mg Each for $100.00
 
Description

Description

Hoechst 33342 is a popular cell-permeant nuclear counterstain that emits blue fluorescence when bound to dsDNA. This dye is often used to distinguish condensed pycnotic nuclei in apoptotic cells and for cell cycle studies in combination with BrdU. It is also available in solution (H-3570).

Apoptosis, Cell Analysis, Cell Cycle, Cell Proliferation, Cell Structure, Cell Tracing & Tracking, Cell Viability & Cytotoxicity, Cell Viability, Proliferation & Function, Cellular Imaging, Chromatin Biology, DNA Fragmentation, Flow Cytometry, Flow Cytometry Apoptosis Assays, Flow Cytometry Cell Cycle Assays, Flow Cytometry Staining by Cell Structure, Flow Cytometry Viability & Cytotoxicity Assays, Flow Cytometry of Cellular Processes, Immunofluorescence (IF), Immunofluorescence Counterstaining, Mounting & Fade Prevention, Immunofluorescence Staining & Detection, Nucleus, Nucleoli & Nuclear Envelope, Organelle Tracing, RNAi, Epigenetics & Non-Coding RNA Research

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Fluorescent
Fluorescence Microscope

For Research Use Only. Not for use in diagnostic procedures.