missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molecular Probes™ FocalCheck™ Microspheres, 6μm DFS Item

Fluorescent green/orange/dark-red ring stains

$411.00 - $420.00

Specifications

Diameter (Metric) 6μm
Format Suspension Beads
Storage Requirements Store in refrigerator (2° to 8°C) and protect from light.
Quantity 0.5mL
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Storage Requirements Price Quantity  
Catalog Number Mfr. No. Storage Requirements Price Quantity  
F14806 Life Technologies
LSF14806
Dark Red, Green, Orange Each for $416.00
F14808 Life Technologies
LSF14808
Blue, Green Each for $411.00
F14807 Life Technologies
LSF14807
Store in refrigerator (2° to 8°C) and protect from light. Each for $420.00
Specifications

Description

The sharp ring stains exhibited by the 6.0μm FocalCheck fluorescent green/orange/dark-red ring stained microspheres produce a striking visual representation of instrument misalignment or other aberrations making them ideal as reference standards for confocal laser-scanning microscopy. Correct image registration is indicated when the multiple ring images are perfectly coincident in all dimensions.

Cell Analysis, Cell Cycle, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF)

Order Info

Shipping Condition: Room temperature

Description & Specifications

Specifications

6μm
Suspension Beads
Store in refrigerator (2° to 8°C) and protect from light.
0.5mL