missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molecular Probes™ FocalCheck™ Microspheres, 15μm DFS Item

Fluorescent green/orange/dark-red ring stains

$426.00 - $433.00

Specifications

Diameter (Metric) 15μm
Format Suspension Beads
Storage Requirements Store in refrigerator (2° to 8°C) and protect from light.
Quantity 0.5mL
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Storage Requirements Price Quantity  
Catalog Number Mfr. No. Storage Requirements Price Quantity  
F7235 Life Technologies
LSF7235
Store in refrigerator (2° to 8°C) and protect from light. Each for $433.00
F7237 Life Technologies
LSF7237
Store in refrigerator (2° to 8°C) and protect from light. Each for $426.00
F7238 Life Technologies
LSF7238
Store in refrigerator (2° to 8°C) and protect from light. Each for $430.00
F7239 Life Technologies
LSF7239
Store in refrigerator (2° to 8°C) and protect from light. Each for $426.00
Specifications

Description

The sharp ring stains exhibited by the 15μm FocalCheck fluorescent green/orange/dark-red ring stained microspheres produce a striking visual representation of instrument misalignment or other aberrations making them ideal as reference standards for confocal laser-scanning microscopy. Correct image registration is indicated when the multiple ring images are perfectly coincident in all dimensions.

Cell Analysis, Cell Cycle, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF)

Order Info

Shipping Condition: Room temperature

Description & Specifications

Specifications

15μm
Suspension Beads
Store in refrigerator (2° to 8°C) and protect from light.
0.5mL