Please login to your online account to display your discounted pricing

Molecular Probes™ Dihydrorhodamine 123 DFS Item

Uncharged and nonfluorescent reactive oxygen species indicator

$281.00 - $284.00

Specifications

Detection Method Fluorescent Dye-Based
Sensor Types Free Radical Sensor
Storage Conditions Store in freezer (-5° to -30°C) and protect from light.
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

D23806

DFS Item
 
life technologies
LSD23806
1mL Each for $281.00

D632

DFS Item
 
life technologies
LSD632
10mg Each for $284.00
Description & Specifications

Specifications

Detection Method Fluorescent Dye-Based
Sensor Types Free Radical Sensor
Storage Conditions Store in freezer (-5° to -30°C) and protect from light.

Dihydrorhodamine 123 can passively diffuse across membranes where it is oxidized to cationic rhodamine 123 which localizes in the mitochondria and exhibits green fluorescence.

Concentration: 5mM

Applications:

Apoptosis, Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Free Radical Detection, Nitro-Oxidative Stress, Reactive Oxygen Species

Shipping Condition: Room Temperature