Please login to your online account to display your discounted pricing

Molecular Probes™ Dihydroethidium (Hydroethidine) DFS Item


Free radical sensor

Manufacturer: life technologies  LSD11347

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. D11347

  • / Each
 

Description & Specifications

Specifications

Storage Conditions Store in freezer (-5° to -30°C) and protect from light.
Quantity 10 x 1mg
Color Orange
Location Cytoplasm and Cytosol, Nucleic Acids
For Use With (Application) Fluorescent Detection