Please login to your online account to display your discounted pricing

Molecular Probes™ DiFMUP (6,8-Difluoro-4-Methylumbelliferyl Phosphate) DFS Item


Continuously assay acid phosphatases at low pH

Manufacturer: life technologies  LSD22065

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. D22065

  • / Each
 

Description & Specifications

Specifications

Color Blue
Target Phosphatases
Storage Requirements Store in freezer (-5° to -30°C).
Quantity 10 x 10mg
Product Type Substrate, Phosphatase
Excitation Wavelength Range 358/455
Dye Type DiFMU
Detection Method Fluorescent
For Use With (Equipment) Microplate Reader