missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molecular Probes™ DiFMUP (6,8-Difluoro-4-Methylumbelliferyl Phosphate) DFS Item

Continuously assay acid phosphatases at low pH

Manufacturer:  Life Technologies LSD6567

 View more versions of this product

Catalog No. D6567


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications Description & Specifications

Specifications

5mg
Fluorescent
Substrate, Phosphatase
DiFMU
Phosphatases
Store in freezer (-5° to -30°C).
Microplate Reader
358/455
Blue