Molecular Probes™ CultureWell™ Chambered Coverglass for Cell Culture DFS Item
Ideal for cell culture and fluorescence imaging applications
$271.00 - $271.00
Specifications
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Products ${productFamilyLength}
Catalog Number Mfr. No. Wells Price Quantity  
Catalog Number Mfr. No. Wells Price Quantity  
C37000 Life Technologies
LSC37000
16 wells per coverglass Each for $271.00
Specifications
Description
The CultureWell removable chambered coverglass provided sterile and ready to use—contains 16 wells that can each hold up to 250µL, allowing cells to be cultured in a number of different conditions on a single slide. When the cells are ready to be imaged, coverglass removal is made easy by the use of a simple tool (Cat. No. C37010, purchased separately) that separates the parts without the need for excessive force, eliminating the risk of coverglass breakage. Frosted microscope slides are also provided for mounting. A sample size containing two chambered coverglasses and the removal tool is also available (Cat. No. C37005)
Cell Analysis, Cell Culture, Cellular Imaging, IHC Sample Preparation, IHC Staining & Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunohistochemistry (IHC), Mammalian Cell Culture
Order Info
Shipping Condition: Room temperature
Description & Specifications
Specifications
0.0