Please login to your online account to display your discounted pricing
Molecular Probes™ CoverWell™ Perfusion Chamber Gasket DFS Item
Designed to facilitate live-cell imaging and manipulation
Manufacturer: life technologies  LSC18120
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. C18120
  • / Each
Description & Specifications
Specifications
Product Type Perfusion Chamber Gasket (1 chamber)
For Use With (Application) Facilitate live-cell imaging and manipulation
Depth (Metric) 0.5mm
Dimensions (L x W) 32 x 19mm