Please login to your online account to display your discounted pricing

Molecular Probes™ CoverWell™ Perfusion Chamber Gasket DFS Item

Designed to facilitate live-cell imaging and manipulation

$151.00 - $263.00

Specifications

For Use With (Application) Facilitate live-cell imaging and manipulation
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Depth (Metric) Diameter (Metric) Dimensions (L x W) Product Type Price Quantity    

C18128

DFS Item
 
life technologies
LSC18128
0.5mm Each for $263.00

C18121

DFS Item
 
life technologies
LSC18121
1.0mm Each for $252.00

C18120

DFS Item
 
life technologies
LSC18120
0.5mm Each for $255.00

C18139

DFS Item
 
life technologies
LSC18139
0.5mm 9mm Each for $151.00
Description & Specifications

Specifications

For Use With (Application) Facilitate live-cell imaging and manipulation

  • CoverWell perfusion chamber gaskets form watertight “press-to-seal” chambers with dual-access ports so reagents can be quickly added and removed
  • Available in single- or multi-well configurations, these gaskets allow multiple experiments to be performed on single microscope slide or coverslip
  • Heat-resistant gaskets can be sterilized and used for direct culturing of cells and tissues

Applications:

Cell Analysis, Cellular Imaging, IHC Sample Preparation, IHC Staining & Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Staining & Detection, Immunohistochemistry (IHC)

Shipping Condition: Room temperature