missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molecular Probes™ CoverWell™ Incubation Chamber Gasket DFS Item

Designed for immunocytochemistry and in situ hybridization

$197.00 - $229.00

Specifications

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Dimensions (L x W) Product Type Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Dimensions (L x W) Product Type Price Quantity  
C18155 Life Technologies
LSC18155
13mm Incubation chamber gasket Each for $198.00
C18151 Life Technologies
LSC18151
40 x 22mm Incubation chamber gasket Each for $229.00
C18150 Life Technologies
LSC18150
40 x 22mm Incubation chamber gasket Each for $197.00
Specifications

Description

These heat resistant, autoclavable and nuclease free chamber gaskets are designed for use with incubation chambers. The chambers improve staining uniformity and sensitivity by enclosing a large sample area while minimizing the reagent volume required. The watertight seal holds reactants in place and prevents evaporation even during prolonged heating and is easily removed for multiple-step procedures.

Applications:

Cell Analysis, Cellular Imaging, IHC Sample Preparation, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunofluorescence Staining and Detection, Immunohistochemistry (IHC)

Order Info

Shipping Condition: Room temperature

Description & Specifications

Specifications

0.0