Please login to your online account to display your discounted pricing
Molecular Probes™ CoverWell™ Imaging Chamber Gasket with Adhesive DFS Item
Incorporate thin, UV-transparent plastic cover, making them ideal for fluorescence microscopy and image analysis
Manufacturer: life technologies  C24726
 View more versions of this product
Catalog No. C24726
  • / Each
Description & Specifications
Specifications
Diameter (Metric) 20mm
Product Type 0.5mm deep gasket
The watertight chambers that form under the covers support and stabilize thick and free-floating specimens, allowing resolution of fine internal structures without the compression or movement artifacts normally seen with ordinary coverslips.
Cell Analysis, Cellular Imaging, IHC Sample Preparation, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunofluorescence Staining and Detection, Immunohistochemistry (IHC)