Please login to your online account to display your discounted pricing
Molecular Probes™ CoverWell™ Imaging Chamber Gasket with Adhesive DFS Item
Incorporate thin, UV-transparent plastic cover, making them ideal for fluorescence microscopy and image analysis
$177.45 - $178.00
Specifications
Diameter (Metric) 20mm
View More Specs
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Products
Catalog Number Mfr. No. Product Type Price Quantity    
C24726
DFS Item
life technologies
LSC24726
0.5mm deep gasket Each for $177.45
C24727
DFS Item
life technologies
LSC24727
1.0mm deep gasket Each for $178.00
Description & Specifications
Specifications
Diameter (Metric) 20mm
The watertight chambers that form under the covers support and stabilize thick and free-floating specimens, allowing resolution of fine internal structures without the compression or movement artifacts normally seen with ordinary coverslips.
Cell Analysis, Cellular Imaging, IHC Sample Preparation, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunofluorescence Staining and Detection, Immunohistochemistry (IHC)
Shipping Condition: Room temperature