Molecular Probes™ CoverWell™ Imaging Chamber Gasket DFS Item
Incorporate thin, UV-transparent plastic cover, making them ideal for fluorescence microscopy and image analysis
$167.00 - $167.00
Specifications
Diameter (Metric) 20mm
View More Specs
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
C18160 Life Technologies
LSC18160
0.5mm deep gasket Each for $167.00
C18161 Life Technologies
LSC18161
1.0mm deep gasket Each for $167.00
Specifications
Description
The watertight chambers form under the covers support and stabilize thick and free-floating specimens, allowing resolution of fine internal structures without the compression or movement artifacts normally seen with ordinary coverslips.
Cell Analysis, Cellular Imaging, IHC Sample Preparation, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunofluorescence Staining and Detection, Immunohistochemistry (IHC)
Order Info
Shipping Condition: Room temperature
Description & Specifications
Specifications
20mm