Please login to your online account to display your discounted pricing
Molecular Probes™ CoverWell™ Imaging Chamber Gasket DFS Item
Incorporate thin, UV-transparent plastic cover, making them ideal for fluorescence microscopy and image analysis
Manufacturer: life technologies  LSC18161
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. C18161
  • / Each
Description & Specifications
Specifications
Product Type 1.0mm deep gasket
Diameter (Metric) 20mm