Please login to your online account to display your discounted pricing

Molecular Probes™ Click-iT™ EdU Alexa Fluor™ 594 HCS Assay DFS Item


Superior alternative to traditional proliferation assays

Manufacturer: life technologies  LSC10355

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. C10355

  • / Each
 

Description & Specifications

Specifications

Color Red
Kit Contents Sufficient material for 10 x 96-well plates
Product Type HCS (High Content Imaging) Assay
Quantity 10 x 96 well plates
Sufficient For 10 x 96 well plates
Detection Method Fluorescent
Dye Type Alexa Fluor 594
Excitation Wavelength Range 590/617
Format 96-well plate
For Use With (Equipment) High Content Instrument
Storage Requirements Store at 2° to 6°C, dessicate and protect from light.