missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molecular Probes™ Click-iT™ EdU Alexa Fluor™ 555 HCS Assay DFS Item

Superior alternative to traditional proliferation assays

Manufacturer:  Life Technologies LSC10352

 View more versions of this product

Catalog No. C10352


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications Description & Specifications

Specifications

2 x 96 well plates
Fluorescent
Sufficient material for 2 x 96-well plates
2 x 96 well plates
555/565
Orange
Store at 2° to 6°C, dessicate and protect from light.
High Content Instrument
Assay, HCS (High Content Imaging)
96-well plate
Alexa Fluor 555