Please login to your online account to display your discounted pricing

Molecular Probes™ CellLight™ Tubulin-RFP, BacMam 2.0 DFS Item


Provides easy method for labeling of tubulin with Red Fluorescent Protein in live cells

Manufacturer: life technologies  LSC10503

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. C10503

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) Confocal Microscope, Floid™ Cell Imaging System, Fluorescence Microscope, Microscope
Detection Method Fluorescent
Label Type RFP (TagRFP)
Color Red-Orange
Wavelength Range 555/584
Quantity 100μL
Target Tubulin