Please login to your online account to display your discounted pricing

Molecular Probes™ CellLight™ Lysosomes-RFP, BacMam 2.0 DFS Item


Provides easy method for labeling of lysosomes with Red Fluorescent Protein in live cells

Manufacturer: life technologies  LSC10504

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. C10504

  • / Each
 

Description & Specifications

Specifications

Wavelength Range 555/584
Label Type RFP (TagRFP)
For Use With (Equipment) Confocal Microscope, Floid™ Cell Imaging System, Fluorescence Microscope, Microscope
Detection Method Fluorescent
Color Red-Orange
Target Lysosomes
Quantity 100μL