Please login to your online account to display your discounted pricing

Molecular Probes™ CellLight™ Lysosomes-RFP, BacMam 2.0 DFS Item


Provides easy method for labeling of lysosomes with Red Fluorescent Protein in live cells

Manufacturer: life technologies  LSC10597

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. C10597

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) Confocal Microscope, Fluorescence Microscope
Detection Method Fluorescent
Label Type RFP (TagRFP)
Color Orange, Red-Orange
Target Lysosomes
Quantity 1mL
Wavelength Range 555/584