Please login to your online account to display your discounted pricing

Molecular Probes™ CellLight™ Lysosomes-GFP, BacMam 2.0 DFS Item


Provides easy method for labeling of lysosomes with Green Fluorescent Protein in live cells

Manufacturer: life technologies  LSC10507

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. C10507

  • / Each
 

Description & Specifications

Specifications

Detection Method Fluorescent
Color Green
Label Type GFP (EmGFP)
Wavelength Range 488/510
For Use With (Equipment) Attune™ Acoustic Focusing Cytometer, Confocal Microscope, Floid™ Cell Imaging System, Fluorescence Microscope, Microscope
Target Lysosomes
Quantity 100μL