Please login to your online account to display your discounted pricing

Molecular Probes™ CellLight™ Actin-GFP, BacMam 2.0 DFS Item


Provides easy method for labeling of actin with Green Fluorescent Protein in live cells

Manufacturer: life technologies  LSC10506

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. C10506

  • / Each
 

Description & Specifications

Specifications

Label Type GFP (EmGFP)
Detection Method Fluorescent
Target Actin
Color Green
Wavelength Range 488/510
For Use With (Equipment) Attune™ Acoustic Focusing Cytometer, Confocal Microscope, Floid™ Cell Imaging System, Fluorescence Microscope, Microscope
Quantity 100μL