Please login to your online account to display your discounted pricing

Molecular Probes™ BackDrop™ Background Suppressor for Microplate Assays DFS Item


Designed to effectively suppress background fluorescence in live cell assays

Manufacturer: life technologies  LSB10512

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. B10512

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Microplate assays, suppress background fluorescence in live cell assays
Includes One 50mL bottle
Quantity 50mL
Quantity 50mL
Storage Requirements Store at 2° to 8°C.