Please login to your online account to display your discounted pricing

Molecular Probes™ BackDrop™ Background Suppressor for Microplate Assays DFS Item


Designed to effectively suppress background fluorescence in live cell assays

Manufacturer: life technologies  LSB10511

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. B10511

  • / Each
 

Description & Specifications

Specifications

Type Signal Enhancers
For Use With (Application) Cellular imaging and microplate assays
Sample Volume 5mL
Quantity 5mL
Description BackDrop™ Background Suppressor, 5mL, 10X Concentration, Red Solution, Signal Enhancers Reagent, Fluorescent Detection Method, Used for Microplate Assays and Cellular Imaging, 2 to 8°C Storage Temperature
Detection Method Fluorescent
Storage Requirements 2 to 8°C
Includes One 5mL bottle
Quantity 5mL