Please login to your online account to display your discounted pricing

Molecular Probes™ aha-dCTP (5-Aminohexylacrylamido-dCTP) DFS Item

Supports two-step technique for improved DNA labeling results

$240.00 - $240.00

Specifications

Product Type Nucleotide, aha-dCTP (5-Aminohexylacrylamido-dCTP)
For Use With (Application) DNA and RNA Purification and Analysis, DNA Labeling, Nucleic Acid Labeling and Oligo Synthesis
Quantity 500μL
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

A32768

DFS Item
 
life technologies
LSA32768
500μL Each for $240.00
Description & Specifications

Specifications

Product Type Nucleotide, aha-dCTP (5-Aminohexylacrylamido-dCTP)
For Use With (Application) DNA and RNA Purification and Analysis, DNA Labeling, Nucleic Acid Labeling and Oligo Synthesis
Quantity 500μL

Aha-dCTP (5-aminohexylacrylamido-dCTP) is modified at the C-5 position of cytosine with a unique hexylacrylamide moiety. aha-dCTP can be used to produce amine-modified DNA by conventional enzymatic incorporation methods such as reverse transcription. The amine-modified DNA can then be labeled with any amine-reactive dye to generate labeled nucleic acid hybridization probes. This two-step labeling technique consistently results in a uniform and high degree of DNA labeling that is difficult to obtain by other methods. In addition to this 2mM solution in TE (500μL unit size), it is available as a 50mM solution in TE (50μL unit size, Cat. No. A32769).

DNA and RNA Purification and Analysis, DNA Labeling, Nucleic Acid Labeling and Oligo Synthesis

Shipping Condition: Wet Ice