missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molecular Probes™ Acridine Orange DFS Item

Cell-permeant nucleic acid binding dye

$84.25 - $134.00

Specifications

Color Orange
Detection Method Fluorescent
For Use With (Equipment) Fluorescence Microscope
Target Nucleic Acids, Nucleus
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Wavelength Range Price Quantity  
Catalog Number Mfr. No. Quantity Wavelength Range Price Quantity  
A1301 Life Technologies
LSA1301
1g 460/650 (RNA), 500/526 (DNA) Each for $134.00
A3568 Life Technologies
LSA3568
10mL 460/650 (RNA) Each for $84.25
Specifications

Description

Acridine Orange emits green fluorescence when bound to dsDNA and red fluorescence when bound to ssDNA or RNA. This unique characteristic makes acridine orange useful for cell-cycle studies. Acridine orange has also been used as a lysosomal dye. This dye is available in solution (A-3568).

Apoptosis, Cell Analysis, Cell Cycle, Cell Structure, Cell Tracing & Tracking, Cell Viability & Cytotoxicity, Cell Viability, Proliferation & Function, DNA Fragmentation, Nucleus, Nucleoli & Nuclear Envelope, Organelle Tracing

Order Info

Shipping Condition: Room Temperature

Description & Specifications

Specifications

Orange
Fluorescent
Fluorescence Microscope
Nucleic Acids, Nucleus